Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hex-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072088.7 | NP_504489.3 | 3 - 4 | 347 | 145 | |
GenBank mRNA | AM748821.2 | - | 3 - 4 | 318 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Caenorhabditis elegans Probe spans exons |