Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pcm-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129372.4 | NP_001122844.1 | 6 - 7 | 685 | 109 | |
NM_072150.7 | NP_504551.3 | 6 - 7 | 749 | 109 | ||
GenBank mRNA | U09669.1 | - | 6 - 7 | 632 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |