Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dhs-16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072153.7 | NP_504554.1 | 1 - 2 | 202 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |