Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acs-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028766.4 | NP_001023937.1 | 3 - 4 | 210 | 76 | |
NM_001028767.7 | NP_001023938.1 | 2 - 3 | 108 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |