Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: oma-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072668.8 | NP_505069.1 | 3 - 4 | 488 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |