Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mdt-27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072985.7 | NP_505386.2 | 3 - 4 | 612 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |