Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: grd-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307786.2 | NP_001294715.1 | 7 - 8 | 1258 | 66 | |
NM_001368461.2 | NP_001355526.1 | 7 - 8 | 1185 | 66 | ||
NM_001368462.3 | NP_001355524.1 | 7 - 8 | 1380 | 66 | ||
NM_001368463.3 | NP_001355525.1 | 8 | 1177 | 66 | ||
NM_001368464.3 | NP_001355528.1 | 7 | 1012 | 66 | ||
NM_001368465.2 | NP_001355527.1 | 7 - 8 | 1039 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Amplicon spans exons and probe does not span exons |