Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hum-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001330875.4 | NP_001317742.1 | 3 - 4 | 743 | 97 | |
NM_001330876.2 | NP_001317741.1 | 3 - 4 | 743 | 97 | ||
NM_001373073.2 | NP_001359965.1 | 4 - 5 | 670 | 97 | ||
NM_001380727.2 | NP_001367720.1 | 3 - 4 | 743 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |