Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: flp-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073043.5 | NP_505444.1 | 4 - 5 | 430 | 80 | |
GenBank mRNA | AF042392.1 | - | 4 - 5 | 427 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |