Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lsy-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269216.3 | NP_001256145.1 | 9 - 10 | 2257 | 66 | |
NM_001269217.3 | NP_001256146.1 | 7 - 8 | 1395 | 66 | ||
NM_001269218.3 | NP_001256147.1 | 10 - 11 | 2382 | 66 | ||
NM_001276918.3 | NP_001263847.1 | 9 - 10 | 2247 | 66 | ||
NM_001276919.3 | NP_001263848.1 | 7 - 8 | 1395 | 66 | ||
NM_001276920.4 | NP_001263849.1 | 10 - 11 | 2382 | 66 | ||
NM_001373080.3 | NP_001359747.1 | 9 | 1616 | 66 | ||
NM_001373081.2 | NP_001360180.1 | 9 | 1597 | 66 | ||
NM_001373082.2 | NP_001359576.1 | 9 | 1616 | 66 | ||
NM_001380733.2 | NP_001368168.1 | 8 - 9 | 1545 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |