Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lsy-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269216.3 | NP_001256145.1 | 2 - 3 | 620 | 119 | |
NM_001269218.3 | NP_001256147.1 | 2 - 3 | 610 | 119 | ||
NM_001276918.3 | NP_001263847.1 | 2 - 3 | 610 | 119 | ||
NM_001276920.4 | NP_001263849.1 | 2 - 3 | 610 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |