Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lea-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269231.3 | NP_001256160.1 | 4 - 5 | 1136 | 75 | |
NM_001269232.3 | NP_001256161.1 | 4 - 5 | 1136 | 75 | ||
NM_001269233.4 | NP_001256162.1 | 4 - 5 | 1136 | 75 | ||
NM_001269234.3 | NP_001256163.1 | 3 - 4 | 1088 | 75 | ||
NM_001269235.3 | NP_001256164.1 | 3 - 4 | 1088 | 75 | ||
NM_001269236.3 | NP_001256165.1 | 3 - 4 | 1088 | 75 | ||
NM_001269240.3 | NP_001256169.1 | 2 - 3 | 872 | 75 | ||
NM_001269241.4 | NP_001256170.1 | 2 - 3 | 872 | 75 | ||
NM_001269242.2 | NP_001256171.1 | 2 - 3 | 890 | 75 | ||
NM_001373085.2 | NP_001360513.1 | 4 | 1136 | 75 | ||
NM_001373086.2 | NP_001360514.1 | 4 | 1136 | 75 | ||
NM_001373087.2 | NP_001360515.1 | 3 | 1088 | 75 | ||
NM_001373088.2 | NP_001360516.1 | 3 - 4 | 1088 | 75 | ||
NM_001373089.2 | NP_001360517.1 | 2 | 872 | 75 | ||
NM_001373090.2 | NP_001360518.1 | 2 | 872 | 75 | ||
GenBank mRNA | AF016513.1 | - | 2 - 3 | 904 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |