Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sas-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269409.2 | NP_001256338.1 | 4 - 5 | 709 | 73 | |
NM_001269410.3 | NP_001256339.1 | 2 - 3 | 290 | 73 | ||
NR_069564.1 | 4 - 5 | 633 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |