Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tre-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269432.3 | NP_001256361.1 | 8 - 9 | 1571 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |