Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: daf-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073559.8 | NP_505960.3 | 5 - 6 | 914 | 112 | |
NR_132532.1 | 4 - 5 | 546 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |