Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hmp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129519.3 | NP_001122991.1 | 5 - 6 | 2408 | 109 | |
NM_001276937.3 | NP_001263866.1 | 5 - 6 | 2407 | 109 | ||
NM_001373121.3 | NP_001360176.1 | 6 | 2414 | 109 | ||
NM_001383414.1 | 5 - 6 | 2408 | 109 | |||
NM_073584.6 | NP_505985.1 | 5 - 6 | 2408 | 109 | ||
GenBank mRNA | AF016852.1 | - | 5 - 6 | 2430 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |