Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073606.5 | NP_506007.1 | 5 - 6 | 658 | 62 | |
GenBank mRNA | AY557405.1 | - | 5 - 6 | 615 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Caenorhabditis elegans Probe spans exons |