Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pkc-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269465.2 | NP_001256394.1 | 7 - 8 | 1320 | 108 | |
NM_001269466.3 | NP_001256395.1 | 11 - 12 | 1716 | 108 | ||
NM_001269467.3 | NP_001256396.1 | 10 - 11 | 1677 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |