Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gpa-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073759.3 | NP_506160.1 | 12 - 13 | 1338 | 147 | |
GenBank mRNA | AY008131.1 | - | 8 - 9 | 907 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |