Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: inft-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073852.8 | NP_506253.4 | 8 - 9 | 2394 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |