Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: uig-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269559.3 | NP_001256488.1 | 5 - 6 | 592 | 131 | |
NM_001269560.3 | NP_001256489.1 | 5 - 6 | 634 | 131 | ||
NM_001380797.1 | NP_001367249.1 | 5 - 6 | 574 | 131 | ||
NM_001380798.1 | NP_001366647.1 | 5 - 6 | 726 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |