Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: uig-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269559.3 | NP_001256488.1 | 8 - 9 | 1567 | 74 | |
NM_001269560.3 | NP_001256489.1 | 8 - 9 | 1609 | 74 | ||
NM_001269563.3 | NP_001256492.1 | 4 - 5 | 1018 | 74 | ||
NM_001269564.4 | NP_001256493.1 | 1 - 2 | 58 | 74 | ||
NM_001380797.1 | NP_001367249.1 | 8 - 9 | 1549 | 74 | ||
NM_001380798.1 | NP_001366647.1 | 8 - 9 | 1701 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |