Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mes-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073932.6 | NP_506333.1 | 5 - 6 | 1450 | 72 | |
GenBank mRNA | AF233290.1 | - | 5 - 6 | 1472 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |