Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: avr-15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028905.8 | NP_001024076.1 | 1 - 2 | 176 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |