Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srxa-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026197.1 | NP_001021368.1 | 3 - 4 | 394 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |