Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074162.3 | NP_506563.2 | 2 - 3 | 748 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |