Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gnrr-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269654.4 | NP_001256583.1 | 5 - 6 | 821 | 80 | |
NM_001269655.4 | NP_001256584.2 | 3 - 4 | 451 | 80 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |