Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nmr-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269711.3 | NP_001256640.1 | 3 - 4 | 575 | 119 | |
GenBank mRNA | AF318614.2 | - | 3 - 4 | 551 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |