Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glb-14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074466.4 | NP_506867.2 | 1 - 2 | 344 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Caenorhabditis elegans Probe spans exons |