Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acr-18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074467.5 | NP_506868.2 | 8 - 9 | 1124 | 80 | |
GenBank mRNA | AY523513.1 | - | 8 - 9 | 1124 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Caenorhabditis elegans Probe spans exons |