Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: twk-34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074505.5 | NP_506906.3 | 9 - 10 | 1580 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |