Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: str-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001322601.3 | NP_001309631.1 | 1 - 2 | 94 | 66 | |
NM_074651.3 | NP_507052.2 | 2 - 3 | 779 | 66 | ||
NR_136372.1 | 3 - 4 | 606 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |