Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: oac-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074713.4 | NP_507114.2 | 4 - 5 | 905 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |