Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srxa-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074718.2 | NP_507119.1 | 1 - 2 | 365 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |