Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ttr-26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_075229.7 | NP_507630.1 | 2 - 3 | 319 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |