Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ttr-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277019.3 | NP_001263948.1 | 2 - 3 | 194 | 105 | |
NM_001277020.2 | NP_001263949.1 | 2 - 3 | 184 | 105 | ||
NM_075574.4 | NP_507975.3 | 2 - 3 | 191 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |