Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: klf-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_075594.4 | NP_507995.2 | 4 - 5 | 779 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |