Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: num-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028926.5 | NP_001024097.1 | 2 - 3 | 477 | 119 | |
NM_001028927.3 | NP_001024098.1 | 3 - 4 | 494 | 119 | ||
NM_001136432.5 | NP_001129904.1 | 5 - 6 | 1450 | 119 | ||
NM_001270029.3 | NP_001256958.1 | 2 - 3 | 452 | 119 | ||
NM_075620.6 | NP_508021.1 | 3 - 4 | 626 | 119 | ||
GenBank mRNA | AF286205.1 | - | 3 - 4 | 626 | 119 | |
AF286206.1 | - | 3 - 4 | 494 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |