Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sel-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_075774.7 | NP_508175.1 | 6 - 7 | 1070 | 69 | |
GenBank mRNA | AF171064.1 | - | 6 - 7 | 1063 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Inventoried Probe spans exons |