Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lim-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270051.2 | NP_001256980.1 | 7 - 8 | 965 | 109 | |
NM_001270052.2 | NP_001256981.1 | 7 - 8 | 723 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |