Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ceh-39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_075941.7 | NP_508342.1 | 3 - 4 | 479 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |