Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gcy-31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029717.2 | NP_001024888.1 | 6 - 7 | 818 | 97 | |
NM_001029718.2 | NP_001024889.1 | 9 - 10 | 1142 | 97 | ||
NM_001029719.1 | NP_001024890.1 | 7 - 8 | 770 | 97 | ||
NM_001330893.1 | NP_001317860.1 | 8 - 9 | 1031 | 97 | ||
GenBank mRNA | AY275182.1 | - | 9 - 10 | 1142 | 97 | |
AY275183.1 | - | 7 - 8 | 770 | 97 | ||
AY275184.1 | - | 6 - 7 | 818 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |