Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: R04B3.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076047.5 | NP_508448.1 | 3 - 4 | 635 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Caenorhabditis elegans Probe spans exons |