Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tag-52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076232.4 | NP_508633.1 | 7 - 8 | 1386 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |