Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tsp-17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136448.4 | NP_001129920.2 | 2 - 3 | 178 | 94 | |
NM_001313384.4 | NP_001300313.1 | 3 | 191 | 94 | ||
NM_001383547.1 | 2 - 3 | 185 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |