Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: abu-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076258.6 | NP_508659.1 | 6 - 7 | 897 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |