Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rab-37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047829.5 | NP_001041294.1 | 3 - 4 | 242 | 160 | |
NM_001313453.4 | NP_001300382.1 | 2 - 3 | 206 | 160 | ||
NM_001373276.3 | NP_001359998.1 | 4 - 5 | 378 | 160 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |