Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acc-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076409.6 | NP_508810.2 | 5 - 6 | 505 | 86 | |
GenBank mRNA | AY849796.1 | - | 5 - 6 | 519 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |