Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hst-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001375033.2 | NP_001362017.1 | 6 - 7 | 858 | 134 | |
GenBank mRNA | AY081844.1 | - | 5 - 6 | 836 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |