Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sulp-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076543.6 | NP_508944.1 | 12 - 13 | 1582 | 101 | |
GenBank mRNA | AY887908.1 | - | 12 - 13 | 1604 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |